Cleanroom sleeve protectors, Tyvek® IsoClean®, model IC501B 00, Non sterile
Leverantör: DuPont
IC501BWH00
DUPOIC501BWH00EA 2100 SEK
DUPOIC501BWH00
Cleanroom sleeve protectors, Tyvek® IsoClean®, model IC501B 00, Non sterile
Engångskläder
Tyvek® IsoClean® sleeve model IC501B. Not clean-processed and not sterilised.
  • Bound internal seams
  • Tunnelled elastication at wrist and bicep
  • Partial body chemical protective clothing
  • Barrier to infective agents
Suitable for use in GMP class C/D (ISO Class 6-9) clean rooms.
Certifikat: Certified according to Regulation (EU) 2016/425; Partial body chemical protective clothing, Category III, Type PB [6-B]; EN 14126 (barrier to infective agents).
Förpackning: Packed in cardboard box with 100 per box, bulk packed. 2 polyethylene liners.