Natriumdeoxicholat
Leverantör: G Biosciences
Varning
Synonyms: Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate
DG091
DG091EA 17650 SEK
DG091
Natriumdeoxicholat
Natriumdeoxicholat
Formel: C₂₄H₃₉NaO₄
MW: 414.56 g/mol
Smältpunkt: 357…365 °C
Storage Temperature: Omgivande
MDL Number: MFCD00064139
CAS nummer: 302-95-4
EINECS: 206-132-7