N,N-Dimetyl-1,4-benzendiamin 2 HCl ≥99.00% suitable for peroxidase analysis, Sigma-Aldrich®

Leverantör: SIGMA ALDRICH MICROSCOPY
Fara

Synonyms: N,N-Dimethyl-p-phenylenediamine dihydrochloride, N,N-Dimethyl-P-phenylenediamine 2HCl

M-Clarity™ quality level = MQ200

D4139-100G D4139-10G D4139-25G
D4139-100GEA 3570 SEK
D4139-100G D4139-10G D4139-25G
N,N-Dimetyl-1,4-benzendiamin 2 HCl ≥99.00% suitable for peroxidase analysis, Sigma-Aldrich®
N,N-Dimetyl-1,4-benzendiamin 2 HCl

N,N-Dimethyl-p-phenylenediamine dihydrochloride (DMPD) has been used in the HID-AB (high iron diamine-alcian blue) staining of intestinal tract samples. N,N-Dimethyl-p-phenylenediamine dihydrochloride (DMPD) is used for the determination of peroxidase. DMPD is converted into a red pigment of semiquinone nature. It is also used in the HID-AB (high iron diamine-alcian blue) staining procedures, which are used to differentiate between sialomucin and sulfomucin in the gastrointestinal tract.


Observera: Darkens readily when exposed to air.

Formel: (CH₃)₂NC₆H₄NH₂·2HCl
MW: 209.12 g/mol
Smältpunkt: 222 °C (dec., lit.)
Storage Temperature: Omgivande
MDL Number: MFCD00012991
CAS nummer: 536-46-9
EINECS: 208-635-7
UN: 2811
ADR: 6.1 II
Merck Index: 13,03280
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f64621224792e0a4614d3b6a46f0a95e29b130a8&vid=f012289c4ede1e0a60734047c7b7b455f6eb867e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Solubility (method) 50 mg/ml in water
Infrared spectrum Conforms to structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR