Supelco® Organic Reference Standard, Citronellol solution
Leverantör: Merck
CRM40469
CRM40469EA 976 SEK
CRM40469
Supelco® Organic Reference Standard, Citronellol solution
Standarder Organic Standards
  • GHS signal: GHS02, GHS06, GHS08
Observera: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311
Order Now
SPECIFIKATIONER
Environmentally Preferable
M-Clarity quality level MQ300
CAS-nr 106-22-9
Faroklassbeskrivning H225 - H301 + H311 + H331 - H370
Flampunkt 9,7 °C - Closed cup
Shipping conditions WICE
Storage conditions 2…8 °C