Supelco® TraceCERT® Organic Reference Standard, Benfuracarb

Leverantör: Merck

CRM06035-100MG
CRM06035-100MGEA 2740 SEK
CRM06035-100MG
Supelco® TraceCERT® Organic Reference Standard, Benfuracarb
Standarder Organic Standards

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. Benfuracarb certified reference material (CRM) may also be used as following:


  • Residue analysis of 18 insecticides in water and sediment samples collected from aquatic areas around the rice paddy fields by chromatography tandem mass spectrometry
  • Analysis of 168 pesticides in paprika using the modified dry ice-partitioning QuEChERS method combined with LC-MS/MS
  • Simultaneous determination of 18 carbamate pesticide residues from whole chestnut and pine nut samples using ultra-performance liquid chromatography coupled with tandem mass spectrometry (UPLC-MS-MS) following a gel permeation chromatography (GPC) clean-up
  • Development and validation of a multi-residue analysis method for detecting 70 pesticides belonging to different classes from drinking water samples using a polymeric sorbent for solid phase extraction (SPE) and gas and liquid chromatography coupled to tandem mass spectrometry (GC-MS/MS and LC-MS/MS) for quantification
  • Detection of 15 organic nitrogen-containing pesticides from vegetable samples using matrix solid-phase dispersion (MSPD) along with gas chromatography-Tandem mass spectrometry (GC-MS-MS)
  • Simultaneous analysis of 27 organophosphorus, organochlorine, pyrethroid, and carbamate pesticides in Radix astragali by microwave-assisted or dispersive-solid phase extraction in combination with GC-MS


Observera: P202 - P261 - P264 - P273 - P301 + P310 - P304 + P340 + P311

Order Now

SPECIFIKATIONER

Environmentally Preferable
M-Clarity quality level MQ300
CAS-nr 82560-54-1
MDL No. MFCD00210247
Molekylvikt 410,53
Faroklassbeskrivning H301 + H331 - H361f - H410
Storage conditions –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR