Self-retracting lifelines, Rebel™, Protecta®, 3M™
Leverantör: CAPITAL SAFETY
3100420 3100423 3590498 3590520 3590521 3590545 3590610
CPSD3100420EA 0 SEK
CPSD3100420 CPSD3100423 115-2678 CPSD3590520 CPSD3590521 CPSD3590545 CPSD3590610
Self-retracting lifelines, Rebel™, Protecta®, 3M™
Fallskydd Fallstopp
These Protecta® Rebel™ Web self-retracting lifelines provide an economical fall protection solution without compromising on performance or safety. They are available in a range of lengths to meet the specific needs of jobsites. Lifelines are available in a super-tough nylon housing comprised of 25 mm polyester webbing, or in a durable, lightweight thermoplastic housing comprised of galvanised steel.
  • Swivelling top anchorage point
  • Self-locking snap hook on lifeline
  • Compact, lightweight design
  • Tough and impact-resistant stackable thermoplastic housing
  • Quick
More Product Information