Small parts pouch, DBI-SALA®
Leverantör: CAPITAL SAFETY
1500119 1500121 1500122 1500123
CPSD1500119EA 0 SEK
CPSD1500119 CPSD1500121 CPSD1500122 CPSD1500123
Small parts pouch, DBI-SALA®
Påsar Pouches and Belt Cases
DBI-SALA® Small Parts Pouch features an innovative self-closure system, designed with drop prevention in mind.
  • Can be used from a belt or harness
  • Made from 18 oz. duck canvas
This pouch makes it nearly impossible for objects to fall out once placed in the bag, while making it easy for retrieval by the user since no opening or closing is necessary. The rear of the small parts pouch features belt loops, and the pouch can also be worn from a harness using a carabiner (not included).