Cleaning swabs, sealed foam, CONSTIX® SF-3
Leverantör: CONTEC
SF-3
CONTSF-3EA 2150 SEK
CONTSF-3
Cleaning swabs, sealed foam, CONSTIX® SF-3
Svabbar Svabbar för rengöring
The SF-3 swab consists of 100 pore/inch sealed closed cell foam head with a PP handle.
  • Swab head is thermally sealed and bonded to the handle without adhesives or binders
  • Large head is ideal for cleaning and applying coatings
The polyurethane foam head provides good particle entrapment and removal.
The SF-3 swab is compatible with many solvents, such as isopropanol and ethanol, but is not recommended for use with ketones, such as acetone or MEK.
Förpackning: Swabs are packed 500 per case.
Observera: *Produkten/produkterna på denna sida liggerMore Product Information