Cleanroom wipes, Anticon® Gold Sorb™
Leverantör: CONTEC
492238-957 492238-956 492238-958 492238-959 492238-955 492238-960
115-0440EA 1275 SEK
115-0440 115-1848 CONT492238-958 CONT492238-959 CONT492238-955 CONT492238-960
Cleanroom wipes, Anticon® Gold Sorb™
Torkdukar Torkdukar, renrum Torr
100% polyester wipe, blend of standard and microfibre filaments with sealed edges.
  • Unique microfibre provides superior residue and particle removal, even when dry
  • Sealed edges to reduce particle and fibre generation
  • High sorptive capacity and fast adsorption rate for superior spill control
  • Manufactured from unique microfibre yarn which provides cleaner performance than other common microfibre wipes
  • Sterile wipes are packaged in low count easy tear bags to reduce waste
Anticon® Gold Sorb™ is a unique wipe which combines the performance of a microfibreMore Product Information