Digi-Sense High-Temperature Thermocouple Probe
Leverantör: CONTROL
08516-96
COCO08516-96EA 0 SEK
COCO08516-96
Digi-Sense High-Temperature Thermocouple Probe
Temperaturgivare
This Digi-Sense Type-K High-Temperature Thermocouple Probe can withstand the harsh demands of industrial environments.
  • Delivers fast, accurate temperature readings
  • Probe is flexible and can be bent around pipe diameters that are twice the probe sheath diameter
  • Response time is 0,85 seconds or greater
  • Braided cable with fiberglass insulation to withstand high temperatures
  • Sheath made from ultra-durable Inconel® 600
  • Includes a color-coded ANSI Type-K mini connector
  • Applications: Industrial environments
The cable features fiberglass insulation, allowingMore Product Information