Hydrogen peroxide test kit, CHEMets®
Leverantör: CHEMetrics
K-5520C
CHETK-5520CEA 2375 SEK
CHETK-5520C
Hydrogen peroxide test kit, CHEMets®
Testkit för miljöanalys
HR CHEMets® is the new way to test at high ranges and replaces the now discontinued VACUettes product line.
Leveransinformation: Test kits (5-50 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): HR CHEMets® refill, flat comparator, 25 ml sample cup, 10 ml syringe, and instructions.