Copper test kit for use with spectrophotometer, Vacu-vials®
Leverantör: CHEMetrics
K-3503
CHETK-3503EA 1650 SEK
CHETK-3503
Copper test kit for use with spectrophotometer, Vacu-vials®
Testkit för miljöanalys
The method measures total soluble copper as ppm (mg/L) Cu. The test kits are applicable for analysis of drinking water, surface waters, groundwater, wastewater and seawater.
  • Premixed
  • Premeasured
  • Precise
Beställningsinformation: This kit requires the use of a CHEMetrics Direct-Readout Photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.
Leveransinformation: Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, 25 mL sample cup, ampoule blank, and instructions.