Vacu-vials® Film Forming Amines Kit
Leverantör: CHEMetrics
K-1013
CHETK-1013EA 2175 SEK
CHETK-1013
Vacu-vials® Film Forming Amines Kit
Testkit för miljöanalys
Film-forming amines, also known as FFAP or FFP, are chemicals applied in high-pressure boilers, cooling systems, and HRSGs to guard against corrosion and reduce the presence of corrosive agents.
  • Analysis time: 2 minutes
The test kit utilises the Rose Bengal method to detect and measure these substances in boiler water or steam condensate.
The kit comes in a cardboard box and contains everything needed to perform 30 tests: 30 ampoules, acidifier solution, 25 ml sample cup, ampoule blank, and instructions.
This kit requires the use of a V-3000 multi-analyte photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell.