Buffer, TBS solution 10X concentrate (TRIS buffered saline) pH 7,4
Leverantör: Cayman Chemical
600232-250 600232-500 600232-1
CAYM600232-250EA 1325 SEK
CAYM600232-250 CAYM600232-500 CAYM600232-1
Buffer, TBS solution 10X concentrate (TRIS buffered saline) pH 7,4
Buffertar Buffertar för elektrofores Tvättbuffertar
Formulation: Concentrated TBS (250 mM Tris-base; pH 7,4; 1,37 M sodium chloride and 27 mM KCl).
Suggested storage (long term) temperature: 4 °C.