Avantor® ACE®, Preparative Column Coupler
Leverantör: Avantor
C0002EA 880 SEK
C0002
Avantor® ACE®, Preparative Column Coupler
Beslag och kopplingar
Column coupler made from stainless steel/PEEK suitable for connecting preparative columns to stand-alone guard cartridge holder, or for making similar HPLC connections.
Ideally suited to connect 30 mm id preparative columns to H0009 cartridge holder.