Syringes, SteriPlast®
Leverantör: BURKLE
5325-0100 5325-0060
BURK5325-0100EA 1225 SEK
BURK5325-0100 BURK5325-0060
Syringes, SteriPlast®
Sprutor Sprutor, medicinska
Transparent PP syringes with printed graduations.
  • Can be extended to any length with additional tubing
  • Luer adapter is supplied for the 100 ml syringe
The syringe is supplied in a sterile package and ready for immediate use.
Order Now
SPECIFIKATIONER
Environmentally Preferable