Melting point apparatus, M-565
Leverantör: BUCHI
11058004
BUCH11058004EA 0 SEK
BUCH11058004
Melting point apparatus, M-565
Smältpunktsapparater
M-565 is an advanced, automated instrument with video capability for professional quality control in a regulated environment. Offers all the features above but with the automated determination of melting and boiling points. The product is available as a 'Professional set' with or without the M-569 sample loader - please see below for details.
  • Real time video displayed in 6× magnification
  • Video replay function with variable speed
  • Exclusive optional accessories including MeltingPoint Monitor software and IQ/OQ documentation
  • Reproducible results with M-569
  • M-5
More Product Information