Electroporation system, exponential decay wave, ECM™ 630
Leverantör: BTX
45-2051 45-0651 45-0652
BTXI45-2051EA 0 SEK
BTXI45-2051 BTXI45-0651 BTXI45-0652
Electroporation system, exponential decay wave, ECM™ 630
Elektroporeringssystem Elektroporatorer
The ECM™ 630 is an exponential decay wave electroporation generator providing a broad range of voltage and time constants for full flexibility in varying applications. The ability to select the resistance and capacitance values and adjust the range of voltages is the key to achieving the optimal time constants and field strengths needed for efficient transformation of prokaryotes and for eukaryote transfection.
  • Wide and accurate exponential decay voltage and pulse length ranges
  • Preset protocols - includes the most common bacteria and microorganism cell
More Product Information