Compact circuit breaker lockout
Leverantör: Brady
836597
BRDY836597EA 215 SEK
BRDY836597
Compact circuit breaker lockout
Säkerhetslås
Compact universal design to lock out a large range of single, double and triple-pole circuit breakers.
Place lockout over circuit breaker handle, tighten set screw and attach padlock and tag.