Magicard™ Colour + Black Film for Magicard™ 300, 250 Shot
Leverantör: Brady
321972
BRDY321972EA 1075 SEK
BRDY321972
Magicard™ Colour + Black Film for Magicard™ 300, 250 Shot
Printer Ribbons
The Magicard™ combined colour/black ribbon is perfect for full colour prints on one side and black prints on the other side, using a Magicard™ 300 printer.
  • High-quality, full colour ribbon for one side with overcoat and black on the other side
  • Includes a cleaning roller for dust free printing
Print 250 cards, full colour on one side with black on the reverse side.
Leveransinformation: Includes 1 dye film and 1 cleaning roller.