Magicard™ Prima 8 Uno Card Printer
Leverantör: Brady
321953
BRDY321953EA 38725 SEK
BRDY321953
Magicard™ Prima 8 Uno Card Printer
Printer Document Printers
The Magicard™ Prima 8 provides a reliable, robust solution for instant issuance and lamination in a compact package.
  • Single-side printing
Using a retransfer printing process, images are first printed onto a thin film then heat transferred onto the card surface. This produces a high-resolution glossy image, printed right over the edge of the card.
Encoding type: None
Visual security: Hologram, UV effect
Certifikat: UKCA.
Beställningsinformation: Warranty: Three-year MagiCover Plus.
Leveransinformation: This Prima 8 printer ships with anMore Product Information