Label printers with peel function and vial label applicator, industrial thermal transfer benchtop, BradyPrinter i7100
Leverantör: Brady
307652 307658 307653 307659
BRDY307652EA 71550 SEK
BRDY307652 116-0938 BRDY307653 BRDY307659
Label printers with peel function and vial label applicator, industrial thermal transfer benchtop, BradyPrinter i7100
Printer Label Printers
The BradyPrinter i7100 industrial thermal transfer benchtop printer is ideal for customers requiring reliable high volume printing on lots of different materials within the aerospace, defence and mass transit, electronics, automotive, logistics, electrical, voice-datacom and LAB markets.
  • Prints labels and then applies them to wires or cables
  • Two step process
  • Laboratory labelling application
  • Lithium-manganese battery
  • Colour: Monocolour
It guarantees accurate printing on sleeves, tags, labels, tapes, cable markers and much more. This printer comes withMore Product Information