T-Flag or P-Flag Matte Polypropylene Label with Ribbon for M510, BMP51, BMP53 and M511 Printers
Leverantör: Brady
170860
BRDY170860EA 745 SEK
BRDY170860
T-Flag or P-Flag Matte Polypropylene Label with Ribbon for M510, BMP51, BMP53 and M511 Printers
Etiketter
White on black B-425 thermal transfer printable polypropylene is ideal for labelling applications requiring excellent solvent resistance and print performance.
  • No visible affect to label when exposed to most common chemicals, oils or water
  • Service temperature of −70 to 80 °C (−94 to 176 °F)
  • Matte finish
Ideal for asset tracking, barcode labelling, component and equipment labelling, data and telecommunications labelling, general identification, inventory and inspection labelling, laboratory labelling, and wire and cable labelling applications.