Electrical plug lockout, 3-in-1
Leverantör: Brady
045842
BRDY045842EA 370 SEK
BRDY045842
Electrical plug lockout, 3-in-1
Säkerhetslås
This electrical plug lockout accomodates high and low-voltage plugs up to 76 mm in diameter and 139 mm in length.
  • Two sliding top lids can be used individually or together to accomodate small, medium and large-diameter cords up to 31 mm
  • Made in bright yellow thermoplastic (PP)
  • With English instruction labels
Max. wire/cable diameter: 31,00 mm
Ø×H: 158,8×88,14 mm