SWIFT OTG Safety Glass
Leverantör: Bollé Safety
SWIOTN10E
BOLESWIOTN10EEA 250 SEK
BOLESWIOTN10E
SWIFT OTG Safety Glass
Skyddsglasögon
SWIFT OTG, the affordable safety over-the-glasses solution designed for superior performance. Equipped with our anti-fog and anti-scratch platinum lite coating, it ensures crystal-clear vision in all situations.
  • Thin, flexible temples with side shields for enhanced comfort and protection
  • Safety glasses with clear lenses are essential indoors
  • Clear lenses provide 90% light transmission
  • Is compatible with the platinum anti-fog coating
  • Indoor use
Its wide, lightweight, and well-balanced frame fits comfortably over most prescription glasses while offeringMore Product Information