Tubing holders, BOLA
Leverantör: Bohlender
F706-06
BOHLF706-06EA 670 SEK
BOHLF706-06
Tubing holders, BOLA
Klämmor Slangklämmor
For tubing with O.D. 1,6 mm (¹/₁₆") and 3,2 mm (¹/₈").
  • Safe fixing of the tubing during the flanging procedure
  • Easy assembly and handling
  • Injuries due to the hot flanging tips are avoided