Electronic overhead stirrer, RS9000
Leverantör: BOCHEM
RS9000
BOCHRS9000EA 27175 SEK
BOCHRS9000
Electronic overhead stirrer, RS9000
Omrörare Omröramotorer
Stirrer for high velocity applications with low viscosity. Laboratory stirrer with variable and constant speed range for media.
  • Digital LCD display
  • Speed range infinitely variable at/with high quiet running
  • Convenient user interface
  • Overload protection and over-heat protection functions
  • Smooth start and change of the stirring speed
  • Impeller can be plugged through
Suitable for most stirring tasks in a laboratory.