Burette and test tube clamps, with bosshead
Leverantör: BOCHEM
5631
BOCH5631EA 360 SEK
BOCH5631
Burette and test tube clamps, with bosshead
Klämmor Universalklämmor
These chromium plated clamps with bosshead (13,5 mm) and PVC coated jaws are designed for holding burettes and test tubes (Ø 7 - 25 mm).