Chairs, Semi Stand-Up and Stools for Cleanrooms
Leverantör: Kango
7EK91GHLE01905 7EK91GHLE00905 7EK90GHLE00905 7EK90GBLN00905 7EK90GBLE00905 7EK91GBLE00905 7EK91GBLN00905 7EK90GHLE01905
BIEN7EK91GHLE01905EA 7000 SEK
BIEN7EK91GHLE01905 BIEN7EK91GHLE00905 BIEN7EK90GHLE00905 BIEN7EK90GBLN00905 BIEN7EK90GBLE00905 BIEN7EK91GBLE00905 BIEN7EK91GBLN00905 BIEN7EK90GHLE01905
Chairs, Semi Stand-Up and Stools for Cleanrooms
Stolar
Complete range of chairs, stools and semi stand-up seats developed for cleanrooms, in PU ESD or vinyl ESD.
  • Does not overheat or cause perspiration
  • Resists sharp tools, sparks, weak acids and hydrocarbons
  • Easily cleaned using common household products
  • Thickness of the foam: 4 cm
Different models and ranges to address all the working positions: Semi stand-up seats, comfort polyurethane chairs or Koncept chairs and stools
Auto-adaptative Koncept range in ESD Polyurethane:
A good working position is a dynamic posture, making it possible to move, becauseMore Product Information