Digital refractometer, RFM700-M
Leverantör: BELLINGHAM STANLEY
19-00 19-10
BELL19-00EA 0 SEK
BELL19-00 BELL19-10
Digital refractometer, RFM700-M
Refraktometrar Refraktometrar, bordsmodeller
Robust fully automatic instruments for the food, sugar and beverage industries where temperature control is not required, such as measuring sugar (°Brix) concentrations in the food processing industry.
  • Automatic sense 'hands free' measurements
  • Simple audit trail with date, time and batch number
  • Alphanumeric keypad for easy data entry
  • Flat sapphire prism surface for easy-cleaning
  • Selectable read type: Continual, single, intelligent auto-sense
Designed with ultimate simplicity in mind, the RFM700-M refractometers featuring automatic temperatureMore Product Information