Soxhlet extractor, multiposition
Leverantör: BEHR
B00218423
BEHRB00218423EA 0 SEK
BEHRB00218423
Soxhlet extractor, multiposition
Utrustning för extraktion Utrustning för Soxhletextraktion
Complete workstation to undertake Soxhlet extraction using 6×100 ml flasks.
Leveransinformation: Workstation comprises temperature and time controller, heating block with six sample positions, condenser frame with coolant water distribution manifold, glassware including flasks, extractors and Dimroth condensers.