Microcentrifuge, refrigerated, Axygen® Axyspin
Leverantör: Corning
601-05-021
AXYG601-05-021EA 72900 SEK
AXYG601-05-021
Microcentrifuge, refrigerated, Axygen® Axyspin
Centrifuger Mikrocentrifuger
Axygen® Axyspin is a high-performance refrigerated microcentrifuge designed to quickly process samples for applications such as nucleic acid and protein preparation, purifications, extractions and pelleting.
  • Powerful refrigeration system cools to 4 °C in eight minutes
  • Exceptionally quiet and compact
Brushless drive technology has been fine-tuned in the microcentrifuge to deliver fast acceleration and deceleration of the solid aluminium rotor. Control panel features a back-lit LCD display, which allows users to easily view running parameters while theMore Product Information