Safranine 0 (3,7-diamino-2,8-dimetyl-5-fenylfenaziniumklorid) Microscopy (C.I. 50240)
Leverantör: PanReac AppliChem
Synonyms: CI 50240
A0427.0100
APPCA0427.0100EA 8225 SEK
APPCA0427.0100
Safranine 0 (3,7-diamino-2,8-dimetyl-5-fenylfenaziniumklorid) Microscopy (C.I. 50240)
Safranine 0 (3,7-diamino-2,8-dimetyl-5-fenylfenaziniumklorid)
Formel: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
Densitet: 1.447 g/cm³ (22 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011759
CAS nummer: 477-73-6
EINECS: 207-518-8