Dehydrated culture media, for microbiology

Leverantör: PanReac AppliChem

413812.1210
APPC413812.1210EA 5275 SEK
APPC413812.1210
Dehydrated culture media, for microbiology
Media Dehydrerade mikrobiologimedia

Culture medium for the detection and enumeration of Enterococci in water and foods.


  • pH: 7,2 ±0,2
  • Soluble in water
  • Store in a cool and dry place


Medium contains yeast extract (5,0 g/l); D(+)-glucose (2,0 g/l); sodium azide (0,4 g/l); dipotassium hydrogen phosphate (4,0 g/l); 2,3,5-triphenyl-2H-tetrazolium chloride (0,1 g/l); tryptose (20,0 g/l) and agar (10,0 g/l).

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR