Tris(1H,1H,5H-octafluoropentyl)phosphate 95%

Leverantör: Apollo Scientific
Varning

Synonyms: Phosphoric Acid Tris(1H,1H,5H-octafluoropentyl) Ester, Tris(2,2,3,3,4,4,5,5-octafluoropentyl) phosphate

PC8751-25G PC8751-5G
APOSPC8751-25GEA 6775 SEK
APOSPC8751-25G APOSPC8751-5G
Tris(1H,1H,5H-octafluoropentyl)phosphate 95%
Tris(1H,1H,5H-octafluoropentyl)phosphate

Mass Spectrometry standard

Formel: C₁₅H₉F₂₄O₄P
MW: 740.17 g/mol
Kokpunkt: 130 °C/0.1mmHg
Flampunkt: >100 °C
Storage Temperature: Omgivande
MDL Number: MFCD00054654
CAS nummer: 355-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR