(3,4,5-Trifluorophenyl)acetic acid 97%

Leverantör: Apollo Scientific
Varning

Synonyms: 3,4,5-Trifluorophenylacetic acid

PC7818H-1G PC7818H-5G PC7818H-10G
APOSPC7818H-1GEA 260 SEK
APOSPC7818H-1G APOSPC7818H-5G APOSPC7818H-10G
(3,4,5-Trifluorophenyl)acetic acid 97%
(3,4,5-Trifluorophenyl)acetic acid
Formel: (C₆H₂F₃)CH₂COOH
MW: 190.12 g/mol
Smältpunkt: 98…101 °C
Storage Temperature: Omgivande
MDL Number: MFCD00083532
CAS nummer: 209991-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR