3-(Trifluoromethyl)crotonic acid 95%

Leverantör: Apollo Scientific
Fara

PC7668-25G PC7668-5G PC7668-100MG
APOSPC7668-25GEA 5725 SEK
APOSPC7668-25G APOSPC7668-5G APOSPC7668-100MG
3-(Trifluoromethyl)crotonic acid 95%
3-(Trifluoromethyl)crotonic acid
Formel: C₅H₅F₃O₂
MW: 154.09 g/mol
Kokpunkt: 158…163 °C
Smältpunkt: 28…30 °C
Densitet: 1.318 g/cm³ (25 °C)
Flampunkt: 79 °C
Storage Temperature: Omgivande
MDL Number: MFCD00014374
CAS nummer: 69056-67-3
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR