4-(Trifluoromethoxy)anisole 98%

Leverantör: Apollo Scientific
Varning

Synonyms: 1-Methoxy-4-(trifluoromethoxy)benzene

PC7434C-100G PC7434C-25G PC7434C-5G
APOSPC7434C-100GEA 1250 SEK
APOSPC7434C-100G APOSPC7434C-25G APOSPC7434C-5G
4-(Trifluoromethoxy)anisole 98%
4-(Trifluoromethoxy)anisole
Formel: C₈H₇F₃O₂
MW: 192.14 g/mol
Kokpunkt: 163…165 °C
Densitet: 1.272 g/cm³ (25 °C)
Flampunkt: 58 °C
Storage Temperature: Omgivande
MDL Number: MFCD00216942
CAS nummer: 710-18-9
EINECS: 615-239-1
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR