2-Chloro-2-(chlorodifluoromethoxy)-1,1,1-trifluoroethane 99+%

Leverantör: Apollo Scientific
Varning

PC56732-5G
APOSPC56732-5GEA 2425 SEK
APOSPC56732-5G
2-Chloro-2-(chlorodifluoromethoxy)-1,1,1-trifluoroethane 99+%
2-Chloro-2-(chlorodifluoromethoxy)-1,1,1-trifluoroethane
Formel: C₃HCl₂F₅O
MW: 218.94 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03093761
CAS nummer: 32778-08-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR