2,4-Difluoro-3-(trifluoromethyl)bromobenzene 98%

Leverantör: Apollo Scientific
Varning

Synonyms: 3-Bromo-2,6-difluorobenzotrifluoride, 1-Bromo-2,4-difluoro-3-(trifluoromethyl)benzene

PC51187-1G PC51187-5G PC51187-10G PC51187-25G
APOSPC51187-1GEA 960 SEK
APOSPC51187-1G APOSPC51187-5G APOSPC51187-10G APOSPC51187-25G
2,4-Difluoro-3-(trifluoromethyl)bromobenzene 98%
2,4-Difluoro-3-(trifluoromethyl)bromobenzene
Formel: C₇H₂BrF₅
MW: 260.99 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18379808
CAS nummer: 1263377-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR