3-Bromo-4-fluorophenylisocyanate

Leverantör: Apollo Scientific
Fara

PC503010-1G PC503010-25G PC503010-5G PC503010-100G
APOSPC503010-1GEA 1025 SEK
APOSPC503010-1G APOSPC503010-25G APOSPC503010-5G APOSPC503010-100G
3-Bromo-4-fluorophenylisocyanate
3-Bromo-4-fluorophenylisocyanate

May form small amount of sediment

Formel: C₇H₃BrFNO
MW: 216.007 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD22557618
CAS nummer: 900574-93-8
UN: 2206
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR