2-Fluoro-4-nitrobenzenesulphonyl chloride

Leverantör: Apollo Scientific
Fara

Synonyms: 2-Fluoro-4-nitrobenzene-1-sulphonyl chloride

PC501516-1G PC501516-250MG PC501516-5G
APOSPC501516-1GEA 1025 SEK
APOSPC501516-1G APOSPC501516-250MG APOSPC501516-5G
2-Fluoro-4-nitrobenzenesulphonyl chloride
2-Fluoro-4-nitrobenzenesulphonyl chloride
Formel: C₆H₃ClFNO₄S
MW: 239.609 g/mol
Kokpunkt: 145 °C
Smältpunkt: 69…70 °C
Storage Temperature: Omgivande
MDL Number: MFCD11858095
CAS nummer: 1146290-36-9
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR