6-Bromo-8-fluoroimidazo[1,2-a]pyridine-2-carboxylic acid

Leverantör: Apollo Scientific
Varning

PC49399-1G PC49399-250MG
APOSPC49399-1GEA 2275 SEK
APOSPC49399-1G APOSPC49399-250MG
6-Bromo-8-fluoroimidazo[1,2-a]pyridine-2-carboxylic acid
6-Bromo-8-fluoroimidazo[1,2-a]pyridine-2-carboxylic acid
Formel: C₈H₄BrFN₂O₂
MW: 259.032 g/mol
Smältpunkt: 240…243 °C
Storage Temperature: Omgivande
MDL Number: MFCD13193299
CAS nummer: 1260784-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR