(2,6-Difluoro-4-methoxyphenyl)acetic acid 95%

Leverantör: Apollo Scientific

PC400721-1G
APOSPC400721-1GEA 990 SEK
APOSPC400721-1G
(2,6-Difluoro-4-methoxyphenyl)acetic acid 95%
(2,6-Difluoro-4-methoxyphenyl)acetic acid
Formel: (C₆H₂F₂OCH₃)CH₂COOH
MW: 186.16 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04115927
CAS nummer: 886498-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR