2-Fluoro-4-hydroxybenzaldehyde 98%

Leverantör: Apollo Scientific
Varning

PC2146-1G PC2146-5G PC2146-10G
APOSPC2146-1GEA 260 SEK
APOSPC2146-1G APOSPC2146-5G APOSPC2146-10G
2-Fluoro-4-hydroxybenzaldehyde 98%
2-Fluoro-4-hydroxybenzaldehyde
Formel: C₇H₅FO₂
MW: 140.11 g/mol
Smältpunkt: 168…170 °C
Storage Temperature: Omgivande
MDL Number: MFCD06797918
CAS nummer: 348-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR