3-Amino-4-fluoro-1H-indazole

Leverantör: Apollo Scientific
Varning

Synonyms: 4-Fluoro-1H-indazol-3-amine

PC200069-5G
APOSPC200069-5GEA 1700 SEK
APOSPC200069-5G
3-Amino-4-fluoro-1H-indazole
3-Amino-4-fluoro-1H-indazole
Formel: C₇H₆FN₃
MW: 151.14 g/mol
Smältpunkt: 172…174 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00119934
CAS nummer: 404827-78-7

Order Now

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR