3-Methyl-5-(propan-2-yloxy)phenol 95%

Leverantör: Apollo Scientific

OR957599-1G
APOSOR957599-1GEA 7450 SEK
APOSOR957599-1G
3-Methyl-5-(propan-2-yloxy)phenol 95%
3-Methyl-5-(propan-2-yloxy)phenol
Formel: C₁₀H₁₄O₂
MW: 166.217 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16997279
CAS nummer: 959112-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR