(S)-2-Isopropylamino-3-methyl-1-butanol 95%

Leverantör: Apollo Scientific

Synonyms: (S)-N-Isopropyl-2-(3-methyl-1-butanol)amine

OR937893-1G
APOSOR937893-1GEA 2025 SEK
APOSOR937893-1G
(S)-2-Isopropylamino-3-methyl-1-butanol 95%
(S)-2-Isopropylamino-3-methyl-1-butanol
Formel: C₈H₁₉NO
MW: 145.25 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00671550
CAS nummer: 112211-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR