1-Boc-3-methylenepyrrolidine 95%

Leverantör: Apollo Scientific
Varning

Synonyms: tert-Butyl 3-methylenepyrrolidine-1-carboxylate, tert-Butyl 3-methylidenepyrrolidine-1-carboxylate

OR914414-5G
APOSOR914414-5GEA 685 SEK
APOSOR914414-5G
1-Boc-3-methylenepyrrolidine 95%
1-Boc-3-methylenepyrrolidine
Formel: C₁₀H₁₇NO₂
MW: 183.25 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD12407903
CAS nummer: 114214-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR