3-Methylpyrazine-2-carboxylic acid

Leverantör: Apollo Scientific
Varning

OR59825-1G
APOSOR59825-1GEA 275 SEK
APOSOR59825-1G
3-Methylpyrazine-2-carboxylic acid
3-Methylpyrazine-2-carboxylic acid
Formel: C6H6N2O2
MW: 138.1200 g/mol
Smältpunkt: 170…177 °C
Storage Temperature: Omgivande
MDL Number: MFCD08705765
CAS nummer: 41110-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR